Learn More
KLRG1 Antibody (8E11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26179-100ul
 View more versions of this product
Product Code. 30024731
  • € 533.00 / 100µL
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KLRG1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA
8E11
Flow Cytometry 1:50-1:200, ELISA
2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
A synthesized peptide derived from Human KLRG1 [UniProt Q96E93]
100 μL
Immunology, Innate Immunity, Signal Transduction
10219
Store at -20 to -70C. Avoid freeze-thaw cycles.