missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Antibody (8E11), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | KLRG1 |
|---|---|
| Clone | 8E11 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029035
|
Novus Biologicals
NBP3-26179-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30024731
|
Novus Biologicals
NBP3-26179-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| KLRG1 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
| A synthesized peptide derived from Human KLRG1 [UniProt Q96E93] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8E11 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10219 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title