Description
- No accelerator to limit type IV allergies.
- Ultra comfortable glove with faster donning thanks to its internal polymer coating.
- Great elasticity and good dexterity.
- Non-slip texture for excellent grip and object manipulation.
- Recommended for medical, industrial, and agri-food environments.
Specifications
Specifications
| Chlorinated | No |
| Accelerator Free | Yes |
| Applications | Industrial, Healthcare, Laboratory |
| AQL | 1.5 |
| Color | Blue |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 3.14 mil |
| Finger Thickness (Metric) | 0.08 mm |
| Latex Free | Yes |
| Show More |