Learn More
Masterflex™ L/S 2-Stop High-Performance Precision Pump Tubing
Excellent biocompatibility that meets multiple classifications
Brand: Masterflex™ 06421-24
Additional Details : Weight : 0.80000kg
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Slightly greater clarity than peroxide-cured tubing
- Smooth surfaces; lower protein binding levels
- Fewer potential leachables
- Pharmaceutical, biotech, peristaltic pumping, and food applications
- Excellent heat resistance, low compression set, odorless, nontoxic, translucent
- For use with Masterflex™ multichannel pump heads
Specifications
2-Stop pump tubing, silicone (platinum-cured), L/S 24 | |
0.38 m | |
-40°C | |
24 (6.4 mm) | |
1.7 bar | |
100°C |
Platinum-Cured Silicone | |
L/S Series Pumps | |
4 | |
1800 mL/min | |
660 mm Hg |