missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06352-100ul
This item is not returnable.
View return policy
Description
L3MBTL2 Polyclonal antibody specifically detects L3MBTL2 in Human samples. It is validated for Western Blot
Specifications
| L3MBTL2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dJ756G23.3, DKFZP761I141, H-l(3)mbt-l, H-l(3)mbt-like protein 2, l(3)mbt-like 2 (Drosophila), L(3)mbt-like protein 2, L3MBT, lethal(3)malignant brain tumor-like protein 2 | |
| Produced in rabbits immunized with E. coli-derived Human L3MBTL2 fragment. | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 83746 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction