missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 220.00 - € 326.00
Specifications
| Antigen | L3MBTL2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680597
|
Novus Biologicals
NBP3-06352-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629677
|
Novus Biologicals
NBP3-06352-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L3MBTL2 Polyclonal antibody specifically detects L3MBTL2 in Human samples. It is validated for Western BlotSpecifications
| L3MBTL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS, pH7.0 | |
| 83746 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dJ756G23.3, DKFZP761I141, H-l(3)mbt-l, H-l(3)mbt-like protein 2, l(3)mbt-like 2 (Drosophila), L(3)mbt-like protein 2, L3MBT, lethal(3)malignant brain tumor-like protein 2 | |
| Produced in rabbits immunized with E. coli-derived Human L3MBTL2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title