missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (IGL/3886R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23945-100ug
This item is not returnable.
View return policy
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for ELISA
Specifications
| Lambda Light Chain | |
| Monoclonal | |
| 0.2 mg/mL | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| IGL/3886R | |
| Unconjugated | |
| IGLC, immunoglobulin lambda constant group | |
| Recombinant full-length human Lambda Light Chain protein | |
| 100 μg | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction