Learn More
Invitrogen™ Lamin B1 Recombinant Rabbit Monoclonal Antibody (10H34L18), Alexa Fluor™ Plus 488, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702972RP488
Description
Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Excitation: 494 nm; Emission: 519 nm; Laser: Blue Laser Filtration: 0.2 μm post-manufacturing filtered.
Lamin B1 is a key member of the nuclear lamina structure, encoded by the LMNB1 gene. It is part of the B-type lamins, ubiquitously expressed across all mammalian cells, providing structural integrity and mechanical support by forming a meshwork along the inner nuclear membrane. Lamin B1 is involved in maintaining the shape and stability of the nucleus, playing a critical role in cellular processes such as genome organization, DNA replication, and transcription regulation. The lamin filaments are type V intermediate filament proteins, enabling the nucleus to withstand mechanical stress and preserve its functionality. Lamin B1's expression and dynamics vary during cell differentiation, influencing gene regulation. Its location within the euchromatin-enriched section of the genome suggests potential roles in chromatin organization and the epithelial-to-mesenchymal transition. Governed by complex interactions with other proteins, lamin B1 is integral to cellular architecture and health, with variations in its levels linked to developmental and disease processes. Recent studies have highlighted the importance of understanding the role and dynamics of lamin B1 in human-induced pluripotent stem cells and germ layer cells, offering insights into differentiation-dependent changes and their implications for nuclear structure.
Specifications
| Lamin B1 | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
| P14733, P20700 | |
| Lmnb1 | |
| Peptides corresponding to human LAMIN B1 [(aa62-aa79) (aa450-aa467)]. | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10H34L18 | |
| Alexa Fluor Plus 488 | |
| Lmnb1 | |
| 0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16906, 4001 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.