Learn More
Invitrogen™ Lamin B1 Recombinant Rabbit Monoclonal Antibody (10H34L18), Alexa Fluor™ Plus 488, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 363.00
Specifications
| Antigen | Lamin B1 |
|---|---|
| Clone | 10H34L18 |
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30131123
|
Invitrogen™
702972RP48820UG |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30132093
|
Invitrogen™
702972RP488 |
50 μL |
€ 363.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Excitation: 494 nm; Emission: 519 nm; Laser: Blue Laser Filtration: 0.2 μm post-manufacturing filtered.
Lamin B1 Recombinant Monoclonal antibody specifically detects Lamin B1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| Lamin B1 | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
| 0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg | |
| Peptides corresponding to human LAMIN B1 [(aa62-aa79) (aa450-aa467)]. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lmnb1 |
| 10H34L18 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Alexa Fluor Plus 488 | |
| Rabbit | |
| Human, Mouse | |
| P14733, P20700 | |
| 16906, 4001 | |
| IgG | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.