Learn More
Laminin gamma 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-87718
 View more versions of this product
Product Code. 18797723
  • € 500.00 / 0.10mL
Description
Laminin gamma 1 Polyclonal specifically detects Laminin gamma 1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Affinity Purified
IgG
Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500