missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (747201) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals MAB43201
This item is not returnable.
View return policy
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Mouse samples. It is validated for Immunocytochemistry.
Specifications
| LAMP-1/CD107a | |
| Monoclonal | |
| 0.81 mg/mL | |
| Immunocytochemistry | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunocytochemistry | |
| 747201 | |
| Unconjugated | |
| PBS with No Preservative | |
| LAMP1 | |
| Partial recombinant human LAMP1/CD107a protein (aa25-370) | |
| 100 μg | |
| Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
| 3916 | |
| Store at -20C in the dark. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction