missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (747201) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 204.00 - € 486.00
Specifications
| Antigen | LAMP-1/CD107a |
|---|---|
| Clone | 747201 |
| Concentration | 0.81 mg/mL |
| Dilution | Immunocytochemistry |
| Applications | Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074736
|
Novus Biologicals
MAB43201-SP |
25 μg |
€ 204.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18081796
|
Novus Biologicals
MAB43201 |
100 μg |
€ 486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Mouse samples. It is validated for Immunocytochemistry.Specifications
| LAMP-1/CD107a | |
| 0.81 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
| PBS with No Preservative | |
| 3916 | |
| Partial recombinant human LAMP1/CD107a protein (aa25-370) | |
| Primary | |
| Store at -20C in the dark. Avoid freeze-thaw cycles. |
| 747201 | |
| Immunocytochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| LAMP1 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title