Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody
The LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody is a key component of the BacMam Histone H3K9me2 Cellullar Assay (A14161).
Brand: Thermo Scientific™ A14156
This item is not returnable.
View return policy
Description
- The LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody is a key component of the BacMam Histone H3K9me2 Cellullar Assay (A14161)
- It allows for the investigation of dimethylation at lysine 9 on histone H3
- This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys9 dimethylation in their natural complexes
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody Allows Users to:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with a homogenous assay format that can be easily miniaturized
- Improve data quality with the advantages of TR-FRET
Specifications
| 1000 nM, 0.15 mg/mL | |
| 100 μg | |
| Liquid | |
| Methylated |
| Tb (Terbium)/Lanthanide Chelates | |
| Store the LanthaScreen™ Tb-anti-histone H3K9me2 Antibody at -20°C | |
| Dry Ice | |
| LanthaScreen |