missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody
The LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody is a key component of the BacMam Histone H3K9me2 Cellullar Assay (A14161).
€ 11580.00 - € 38540.00
Specifications
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
|---|---|
| Concentration | 1000 nM, 0.15 mg/mL |
| Content And Storage | Store the LanthaScreen™ Tb-anti-histone H3K9me2 Antibody at -20°C |
| Form | Liquid |
| Product Line | LanthaScreen |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
12887542
|
Thermo Scientific™
A14156 |
100 μg |
€ 11580.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
12877542
|
Thermo Scientific™
A14151 |
1 mg |
€ 38540.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody is a key component of the BacMam Histone H3K9me2 Cellullar Assay (A14161)
- It allows for the investigation of dimethylation at lysine 9 on histone H3
- This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys9 dimethylation in their natural complexes
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K9me2 Antibody Allows Users to:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with a homogenous assay format that can be easily miniaturized
- Improve data quality with the advantages of TR-FRET
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Store the LanthaScreen™ Tb-anti-histone H3K9me2 Antibody at -20°C | |
| LanthaScreen | |
| Methylated |
| 1000 nM, 0.15 mg/mL | |
| Liquid | |
| Dry Ice |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title