Learn More
Invitrogen™ LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
Brand: Invitrogen™ PV4756
Description
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
For use in combination with the GFP-4E-BP1 physiological protein substrate (Cat. No. PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled p4E-BP1 (pThr46) antibody in HEPES buffered saline. Store at -20°C. |
|
| Human | |
| End Point | |
| Ser/Thr Kinases |
| 1 mg | |
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| LanthaScreen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.