Learn More
Invitrogen™ LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
€ 4515.00 - € 21780.00
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10297024
|
Invitrogen™
PV4755 |
25 μg |
€ 4515.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10567774
|
Invitrogen™
PV4756 |
1 mg |
€ 21780.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
For use in combination with the GFP-4E-BP1 physiological protein substrate (Cat. No. PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications
| Tb (Terbium)/Lanthanide Chelates |