Learn More
Invitrogen™ LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
€ 4515.00 - € 21780.00
Specifications
Conjugate Tb (Terbium)/Lanthanide Chelates
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10297024
View Documents
Invitrogen™
PV4755
25 μg
€ 4515.00
25µg
View alternative products
Save Up to 
»
10567774
View Documents
Invitrogen™
PV4756
1 mg
€ 21780.00
1mg
View alternative products
Save Up to 
»
Description
Description
A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.
For use in combination with the GFP-4E-BP1 physiological protein substrate (Cat. No. PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications
Specifications
Tb (Terbium)/Lanthanide Chelates
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.