missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib-13C2,15N, TRC

Product Code. 30474685
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474685 5 mg 5mg
30481643 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30474685 Supplier Toronto Research Chemicals Supplier No. TRCL1758045MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246819-07-7
Molecular Formula C2713C2H26ClFN315NO4S
Molecular Weight (g/mol) 584.04
Synonym N-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinamine-13C2,15N, GSK 572016-13C2,15N, GW 572016-13C2,15N, GW 572016X-13C2,15N
SMILES FC1=CC(COC(C(Cl)=C2)=CC=C2NC3=NC=NC4=CC=C(C5=CC=C(C[15NH][13CH2][13CH2]S(C)(=O)=O)O5)C=C43)=CC=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H26ClFN4O4S/c1-40(36,37)12-11-32-16-23-7-10-27(39-23)20-5-8-26-24(14-20)29(34-18-33-26)35-22-6-9-28(25(30)15-22)38-17-19-3-2-4-21(31)13-19/h2-10,13-15,18,32H,11-12,16-17H2,1H3,(H,33,34,35)/i11+1,12+1,32+1
Chemical Name or Material Lapatinib-13C2,15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.