missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lass5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | Lass5 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230279
|
Novus Biologicals
NBP3-35736-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229429
|
Novus Biologicals
NBP3-35736-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lass5 Polyclonal antibody specifically detects Lass5 in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Lass5 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| CerS5, FLJ25304, LAG1 homolog, ceramide synthase 5, LAG1 longevity assurance homolog 5, LAG1 longevity assurance homolog 5 (S. cerevisiae), MGC45411, Trh4 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Lass5 (NP_001268660.1).,, Sequence:, MMKPRPKRFIAKPCALCIGIEDSGPYQAQPNAILEKVFISITKYPDKKRLEGLSKQLDWNVRKIQCWFRHRRNQDKPPTLTKFCESMWRFTFYLCIFCYG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 91012 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title