missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCN8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145410
This item is not returnable.
View return policy
Description
May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.
Specifications
| LCN8 | |
| Polyclonal | |
| Unconjugated | |
| LCN8 | |
| 9230106L18Rik; EP17; epididymal 17 kDa lipocalin; Epididymal-specific lipocalin-8; LCN5; LCN8; lipocalin 5; lipocalin 8; mEP17 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 78076 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q924P3 | |
| LCN8 | |
| Recombinant Mouse LCN8 protein, Leu27-Val176 (Accession Q924P3). | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction