missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCN8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 185.00 - € 405.00
Specifications
| Antigen | LCN8 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17990453
![]() |
Invitrogen™
PA5145409 |
200 μL |
€ 405.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17958782
|
Invitrogen™
PA5145410 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.Specifica
| LCN8 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q924P3 | |
| 78076 | |
| Recombinant Mouse LCN8 protein, Leu27-Val176 (Accession Q924P3). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 9230106L18Rik; EP17; epididymal 17 kDa lipocalin; Epididymal-specific lipocalin-8; LCN5; LCN8; lipocalin 5; lipocalin 8; mEP17 | |
| LCN8 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| LCN8 |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto