missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LCN8 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145409

 View more versions of this product

Product Code. 17990453

  • € 405.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LCN8
Polyclonal
Unconjugated
LCN8
9230106L18Rik; EP17; epididymal 17 kDa lipocalin; Epididymal-specific lipocalin-8; LCN5; LCN8; lipocalin 5; lipocalin 8; mEP17
Rabbit
Antigen affinity chromatography, Protein A
RUO
78076
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Q924P3
LCN8
Recombinant Mouse LCN8 protein, Leu27-Val176 (Accession Q924P3).
200 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.