missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Chromate, TRC

Product Code. 30466014
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466014 10 g 10g
30481177 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30466014 Supplier Toronto Research Chemicals Supplier No. TRCL32000510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-97-6
Molecular Formula CrO4Pb
Molecular Weight (g/mol) 323.19
Synonym Lead Chromate(VI) (PbCrO4), Chromium Lead Oxide (CrPbO4), Lead ChromateLead Chromate (PbCrO4), Lead Chromium Oxide (PbCrO4), Plumbous ChromateRoyal Yellow 6000
IUPAC Name dioxido(dioxo)chromium;lead(2+)
SMILES [Pb+2].[O-][Cr](=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/Cr.4O.Pb/q;;;2*-1;+2
Formula Weight 323.8968
Chemical Name or Material Lead(II) Chromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.