missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Pb/Mg, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Lead, Pb/Mg, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Aluminium |
| CAS | 7439-92-1 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.023 %/Aluminium <0.001 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction