missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Residuals, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Residuals, 50g
Specifications
Specifications
| Chemical Name or Material | Bismuth/Cadmium/Copper/Iron/Nickel/Silver/Tin |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction