missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Residuals, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Residuals, 50g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Nickel/Silver/Thallium/Arsenic/Bismuth/Copper/Tin/Antimony/Selenium/Iron |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0008 %/Nickel 0.0011 %/Silver 0.003 %/Thallium 0.0042 %/Arsenic 0.0095 %/Bismuth 0.0117 %/Copper 0.0551 %/Tin 0.119 %/Antimony 0.497 %/Selenium (0.0001) %/Iron (0.0003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction