missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Residuals, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Residuals, 50g
Specifications
Specifications
| Chemical Name or Material | Sulfur/Magnesium/Sodium/Nickel/Aluminium/Iron/Calcium/Selenium/Zinc/Gold/Tellurium/Indium/Antimony/Tin/Arsenic/Copper/Bismuth/Cadmium/Silver/Lead |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Sulfur -10 ppm/Magnesium -2 ppm/Sodium -2 ppm/Nickel 0.0002 %/Aluminium 0.00021 %/Iron 0.0003 %/Calcium 0.00034 %/Selenium 0.001 %/Zinc 0.0011 %/Gold 0.0015 %/Tellurium 0.0062 %/Indium 0.0069 %/Antimony 0.0116 %/Tin 0.015 %/Arsenic 0.04 %/Copper 0.046 %/Bismuth 0.049 %/Cadmium 0.068 %/Silver 0.093 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction