missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN-28A Antibody (14E6-4E6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22481-0.025mg
This item is not returnable.
View return policy
Description
LIN-28A Monoclonal specifically detects LIN-28A in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| LIN-28A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 5 μg, Immunohistochemistry-Paraffin 1:20 - 1:200, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| LIN28A | |
| Full-length human recombinant protein expressed in bacteria | |
| 0.025 mg | |
| Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells | |
| 79727 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 14E6-4E6 | |
| Unconjugated | |
| CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction