missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN-28A Antibody (14E6-4E6), Novus Biologicals™
Mouse Monoclonal Antibody
€ 200.00
Specifications
| Antigen | LIN-28A |
|---|---|
| Clone | 14E6-4E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 5 μg, Immunohistochemistry-Paraffin 1:20 - 1:200, Chromatin Immunoprecipitation (ChIP) 1-3 ul |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
LIN-28A Monoclonal specifically detects LIN-28A in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| LIN-28A | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells | |
| CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
| LIN28A | |
| IgG2a | |
| Protein A purified |
| 14E6-4E6 | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 5 μg, Immunohistochemistry-Paraffin 1:20 - 1:200, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 79727 | |
| Full-length human recombinant protein expressed in bacteria | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title