Learn More
Lipocalin-2/NGAL Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30700-1-AP-20UL
 View more versions of this product
Product Code. 18030636
  • € 145.00 / 20µL
Description
The Lipocalin-2/NGAL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Lipocalin-2/NGAL. This antibody recognizes human, mouse antigen. The Lipocalin-2/NGAL antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Lipocalin-2/NGAL
200 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
24p3, HNL, LCN2, lipocalin 2, NGAL, Oncogene 24p3, p25
20 μL
3934
Store at -20°C. Stable for one year after shipment.