Learn More
Lipocalin-2/NGAL Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30700-1-AP-20UL
Description
The Lipocalin-2/NGAL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Lipocalin-2/NGAL. This antibody recognizes human, mouse antigen. The Lipocalin-2/NGAL antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Lipocalin-2/NGAL | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| 24p3, HNL, LCN2, lipocalin 2, NGAL, Oncogene 24p3, p25 | |
| 20 μL | |
| 3934 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |