Learn More
Lipocalin-2/NGAL Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | Lipocalin-2/NGAL |
|---|---|
| Concentration | 200 μg/mL |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030636
|
Proteintech
30700-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18025185
|
Proteintech
30700-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Lipocalin-2/NGAL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Lipocalin-2/NGAL. This antibody recognizes human, mouse antigen. The Lipocalin-2/NGAL antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Lipocalin-2/NGAL | |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| Rabbit | |
| 3934 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 200 μg/mL | |
| Liquid | |
| Human, Mouse | |
| 24p3, HNL, LCN2, lipocalin 2, NGAL, Oncogene 24p3, p25 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.