missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipoprotein a Antibody (4H1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NBP2-37477-0.025ml
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Lipoprotein a Monoclonal specifically detects Lipoprotein a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| Lipoprotein a | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| AK38, antiangiogenic AK38 protein, apo(a), APOA, apolipoprotein(a), EC 3.4.21, EC 3.4.21.-, EC 3.4.21.7, lipoprotein, Lp(a), LP, Lp(a) | |
| Mouse | |
| 501 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4H1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| LPA | |
| Purified recombinant fragment of human LPA (AA: 4330-4521) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 4018 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto