missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipoprotein a Antibody (4H1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
€ 196.00 - € 608.00
Specifications
| Antigen | Lipoprotein a |
|---|---|
| Clone | 4H1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620176
|
Novus Biologicals
NBP2-37477-0.025ml |
0.025 mL |
€ 196.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18746743
|
Novus Biologicals
NBP2-37477 |
0.1 mL |
€ 608.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipoprotein a Monoclonal specifically detects Lipoprotein a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Lipoprotein a | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 4018 | |
| Purified recombinant fragment of human LPA (AA: 4330-4521) expressed in E. Coli. | |
| Primary | |
| 501 kDa |
| 4H1 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Mouse | |
| Human | |
| AK38, antiangiogenic AK38 protein, apo(a), APOA, apolipoprotein(a), EC 3.4.21, EC 3.4.21.-, EC 3.4.21.7, lipoprotein, Lp(a), LP, Lp(a) | |
| LPA | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title