missing translation for 'onlineSavingsMsg'
Learn More

Lodenafil Carbonate, TRC

Product Code. 17952985
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952985 10 mg 10mg
30137061 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17952985 Supplier Toronto Research Chemicals Supplier No. TRCL46930010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 398507-55-6
InChI Formula InChI=1S/C47H62N12O11S2/c1-7-11-35-39-41(54(5)52-35)45(60)50-43(48-39)33-29-31(13-15-37(33)67-9-3)71(63,64)58-21-17-56(18-22-58)25-27-69-47(62)70-28-26-57-19-23-59(24-20-57)72(65,66)32-14-16-38(68-10-4)34(30-32)44-49-40-36(12-8-2)53-55(6)42(40)46(61)51-44/h13-16,29-30H,7-12,17-28H2,1-6H3,(H,48,50,60)(H,49,51,61)
IUPAC Name bis[2-[4-[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]sulfonylpiperazin-1-yl]ethyl] carbonate
Molecular Formula C47 H62 N12 O11 S2
Molecular Weight (g/mol) 1035.2
Chemical Name or Material Lodenafil Carbonate

Specifications

Quantity 10 mg
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5,5'-[carbonylbis[oxy-2,1-ethanediyl-4,1-piperazinediylsulfonyl(6-ethoxy-3,1-phenylene)]]bis[1,6-dihydro-1-methyl-3-propyl-, 1-Piperazineethanol, 4-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-, carbonate (2:1) (ester) (9CI), Lodenafil carbonate
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CCOC(=O)OCCN5CCN(CC5)S(=O)(=O)c6ccc(OCC)c(c6)C7=Nc8c(CCC)nn(C)c8C(=O)N7)CC4
Formula Weight 1034.4102
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.