Learn More
Lomerizine Dihydrochloride, TRC
Product Code. 30133933
Change view
Click to view available options
Quantity:
10 mg
250 mg
500 mg
Unit Size:
10mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133933 500 mg 500mg
30137753 250 mg 250mg
30005598 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133933 Supplier Toronto Research Chemicals Supplier No. TRCL469430500MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101477-54-7
Molecular Formula C27 H30 F2 N2 O3 . 2 Cl H
Molecular Weight (g/mol) 541.46
Synonym Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]-, hydrochloride (1:2) (ACI), Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]-, dihydrochloride (9CI), KB 2796, Lomerizine dihydrochloride, Lomerizine hydrochloride, 1-[Bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine hydrochloride (1:2)
IUPAC Name 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride
SMILES Cl.Cl.COc1ccc(CN2CCN(CC2)C(c3ccc(F)cc3)c4ccc(F)cc4)c(OC)c1OC
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C27H30F2N2O3.2ClH/c1-32-24-13-8-21(26(33-2)27(24)34-3)18-30-14-16-31(17-15-30)25(19-4-9-22(28)10-5-19)20-6-11-23(29)12-7-20;;/h4-13,25H,14-18H2,1-3H3;2*1H
Formula Weight 540.1758
Chemical Name or Material Lomerizine, Dihydrochloride