missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Low Alloy, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
CASRegistryNumber1t Iron, Low Alloy, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Zirconium/Lead/Boron/Zinc/Tellurium/Sulfur/Phosphorus/Tin/Arsenic/Aluminium/Niobium/Titanium/Antimony/Molybdenum/Cobalt/Tungsten/Copper/Vanadium/Silicon/Chromium/Manganese/Nickel/Carbon/Nitrogen/Oxygen/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.0017 %/Lead 0.0023 %/Boron 0.0047 %/Zinc 0.0091 %/Tellurium 0.0097 %/Sulfur 0.02 %/Phosphorus 0.045 %/Tin 0.047 %/Arsenic 0.053 %/Aluminium 0.072 %/Niobium 0.077 %/Titanium 0.116 %/Antimony 0.145 %/Molybdenum 0.166 %/Cobalt 0.169 %/Tungsten 0.31 %/Copper 0.433 %/Vanadium 0.437 %/Silicon 1.39 %/Chromium 1.48 %/Manganese 1.87 %/Nickel 2.66 %/Carbon 3.03 %/Nitrogen (<150) ppm/Oxygen (<50) ppm/Iron (<89) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction