missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRCH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15800-1-AP-150UL
This item is not returnable.
View return policy
Description
LRCH3 (Leucine Rich Repeats And Calponin Homology Domain Containing 3) is a Protein Coding gene.Specifications
| LRCH3 | |
| Polyclonal | |
| Unconjugated | |
| LRCH3 | |
| LRCH3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303883, 70144, 84859 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BVU0, Q96II8 | |
| Lrch3 | |
| LRCH3 Fusion Protein Ag8511 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction