missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRCH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | LRCH3 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16855144
|
Proteintech
15800-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16845144
|
Proteintech
15800-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRCH3 (Leucine Rich Repeats And Calponin Homology Domain Containing 3) is a Protein Coding gene.Specifications
| LRCH3 | |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8BVU0, Q96II8 | |
| 303883, 70144, 84859 | |
| LRCH3 Fusion Protein Ag8511 | |
| Primary | |
| -20°C | |
| LRCH3 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| LRCH3 | |
| Lrch3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title