Learn More
LRP2 Antibody (10B5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26163-100ul
 View more versions of this product
Product Code. 30027949
  • € 533.00 / 100µL
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRP2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA
10B5
Flow Cytometry 1:50-1:200, ELISA
calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin
A synthesized peptide derived from Human LRP2 [UniProt P98164]
100 μL
Lipid and Metabolism
4036
Store at -20 to -70C. Avoid freeze-thaw cycles.