missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody (10B5), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | LRP2 |
|---|---|
| Clone | 10B5 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029783
|
Novus Biologicals
NBP3-26163-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30027949
|
Novus Biologicals
NBP3-26163-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| LRP2 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| A synthesized peptide derived from Human LRP2 [UniProt P98164] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 10B5 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 4036 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title