missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody (545818), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB95781
This item is not returnable.
View return policy
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LRP2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| Mouse myeloma cell line NS0-derived partial recombinant human Megalin/LRP2 protein (amino acids 3510-3964) | |
| 100 μg | |
| Lipid and Metabolism | |
| 4036 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 545818 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction