missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody (545818), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
€ 229.00 - € 494.00
Specifications
| Antigen | LRP2 |
|---|---|
| Clone | 545818 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649894
|
Novus Biologicals
MAB95781 |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619804
|
Novus Biologicals
MAB95781-SP |
25 μg |
€ 229.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| LRP2 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| PBS | |
| 4036 | |
| IgG1 | |
| Protein A or G purified |
| 545818 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| Mouse myeloma cell line NS0-derived partial recombinant human Megalin/LRP2 protein (amino acids 3510-3964) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title