Learn More
Invitrogen™ LRRN1 Monoclonal Antibody (K16288_6D3), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | LRRN1 |
|---|---|
| Clone | K16288_6D3 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203525
|
Invitrogen™
MA556210 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30203847
|
Invitrogen™
MA555830 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: ECPQLCVCEI RPWFTPQSTY REATTVDCND LRLTRIPSNL SSDTQVLLLQ SNNIAKTVDE LQQLFNLTEL DFSQNNFTNI KEVGLANLTQ LTTLHLEENQ ITEMTDYCLQ DLSNLQELYI NHNQISTISA HAFAGLKNLL RLHLNSNKLK VIDSRWFDST PNLEILMIGE NPVIGILDMN FKPLANLRSL VLAGMYLTDI PGNALVGLDS LESLSFYDNK LVKVPQLALQ KVPNLKFLDL NKNPIHKIQE GDFKNMLRLK ELGINNMGEL VSVDRYALDN LPELTKLEAT NNPKLSYIHR LAFRSVPALE SLMLNNNALN AIYQKTVESL PNLREISIHS NPLRCDCVIH WINSNKTNIR FMEPLSMFCA MPPEYKGHQV KEVLIQDSSE QCLPMISHDS FPNRLNVDIG TTVFLDCRAM AEPEPEIYWV TPIGNKITVE TLSDKYKLSS EGTLEISNIQ IEDSGRYTCV AQNVQGADTR VATIKVNGTL LDGTQVLKIY VKQTESHSIL VSWKVNSNVM TSNLKWSSAT MKIDNPHITY TARVPVDVHE YNLTHLQPST DYEVCLTVSN IHQQTQKSCV NVTTKNAAFA VDISDQETST
LRRN1 Monoclonal antibody specifically detects LRRN1 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| LRRN1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2810047E21Rik; fibronectin type III, immunoglobulin and leucine rich repeat domains 3; FIGLER3; Kiaa1497; leucine rich repeat neuronal 1; leucine rich repeat protein 1, neuronal; leucine-rich repeat neuronal protein 1; LRN; Lrrn1; Nbla10449; Neuronal leucine-rich repeat protein 1; NLRR-1; UNQ693/PRO1338 | |
| Recombinant human LRRN1 (immunogen range is 31-630). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LRRN1 |
| K16288_6D3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6UXK5 | |
| 57633 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.