missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat anti-Mouse, BB700, Clone: 1A8, BD Horizon™
Rat Monoclonal Antibody
€ 146.94 - € 392.98
Specifications
| Antigen | Ly-6G |
|---|---|
| Clone | 1A8 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15805069
|
BD Biosciences
566453 |
25 μg |
€ 151.00 € 146.94 / Each Save € 4.06 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15875049
|
BD Biosciences
566435 |
0.1 mg |
€ 395.00 € 392.98 / Each Save € 2.02 0.51% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly-6G Monoclonal antibody specifically detects Ly-6G in Mouse samples. It is validated for Flow Cytometry.Specifications
| Ly-6G | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
| 1A8 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Rat | |
| Mouse | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title