missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lyb-2.1 Monoclonal Antibody (CT-72.1), Biotin

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA517925

 View more versions of this product

Product Code. 15334201

  • € 336.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lyb-2.1 Monoclonal Antibody for Flow

CD72.1 (Lyb-2.1), a type II integral membrane glycoprotein and a member of the C-lectin family of cell surface receptors, is a differentiation antigen of B cells and is found in mouse strains expressing the Lyb-2.1 allotype. CD72.1 is the ligand of CD5 which is distributed on all T cells and a small number of B cells. The 10.1.D2 monoclonal antibody blocks binding of CD5 to CD72 on the B cell surface which leads to inhibition of the positive signal resulting from CD5/CD72 pairing. However, 10.1.D2 can itself trigger a positive signal by binding CD72.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lyb-2.1
Monoclonal
0.1 mg/mL
PBS with 0.02% sodium azide
P21855
Cd72
Mouse CD72.1.
100 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry
CT-72.1
Biotin
Cd72
B-cell differentiation antigen CD72; CD72; CD72 antigen; CD72c; Ly-19; Ly32; Ly-32; Lyb-2; Ly-m19; lymphocyte antigen 32
Mouse
Protein G
RUO
12517
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.