missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lyb-2.1 Monoclonal Antibody (CT-72.1), Biotin

Mouse Monoclonal Antibody

€ 351.00 - € 640.00

Specifications

Antigen Lyb-2.1
Clone CT-72.1
Concentration 0.1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15334201
View Documents
Invitrogen™
MA517925
100 μg
€ 351.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15344201
View Documents
Invitrogen™
MA517926
300 μg
€ 640.00
300µg
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Descrizione

Descrizione

Lyb-2.1 Monoclonal Antibody for Flow

CD72.1 (Lyb-2.1), a type II integral membrane glycoprotein and a member of the C-lectin family of cell surface receptors, is a differentiation antigen of B cells and is found in mouse strains expressing the Lyb-2.1 allotype. CD72.1 is the ligand of CD5 which is distributed on all T cells and a small number of B cells. The 10.1.D2 monoclonal antibody blocks binding of CD5 to CD72 on the B cell surface which leads to inhibition of the positive signal resulting from CD5/CD72 pairing. However, 10.1.D2 can itself trigger a positive signal by binding CD72.
Specifica

Specifica

Lyb-2.1
0.1 mg/mL
Flow Cytometry
Biotin
Mouse
RUO
PBS with 0.02% sodium azide
P21855
12517
Mouse CD72.1.
Antibody
CT-72.1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG2a
Mouse
Cd72
B-cell differentiation antigen CD72; CD72; CD72 antigen; CD72c; Ly-19; Ly32; Ly-32; Lyb-2; Ly-m19; lymphocyte antigen 32
Cd72
Primary
Protein G
Videos
SDS
Documenti

Documenti

Certificati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato