missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYRM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54901
This item is not returnable.
View return policy
Description
LYRM1 Polyclonal specifically detects LYRM1 in Human samples. It is validated for Western Blot.
Specifications
| LYRM1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| A211C6.1, LYR motif containing 1, LYR motif-containing protein 1 | |
| Rabbit | |
| 14 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O43325 | |
| LYRM1 | |
| Synthetic peptides corresponding to LYRM1(LYR motif containing 1) The peptide sequence was selected from the middle region of LYRM1. Peptide sequence KEKQYILNEARTLFRKNKNLTDTDLIKQCIDECTARIEIGLHYKIPYPRP. | |
| Affinity purified | |
| RUO | |
| 57149 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction