Learn More
M-CSFR/CD115 Antibody (AFS98) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-43363-0.1mg
 View more versions of this product
Product Code. 18730714
  • € 550.00 / 0.10mg
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
M-CSF R/CD115
Monoclonal
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry < 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing
P07333
CSF1R
The immunogen for this antibody was CD115.
0.1 mg
Myeloid derived Suppressor Cell, Protein Kinase
1436
Store at 4C. Do not freeze.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Blocking Assay
AFS98
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Rat
Affinity Purified
RUO
Primary
Mouse
IgG2a κ