missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-CSFR/CD115 Antibody (AFS98) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
€ 204.00 - € 795.00
Specifications
| Antigen | M-CSF R/CD115 |
|---|---|
| Clone | AFS98 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry < 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing |
| Applications | Blocking Assay, Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249163
|
Novus Biologicals
NBP1-43363-0.025MG |
0.025 mg |
€ 204.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18272694
|
Novus Biologicals
NBP1-43363-0.05MG |
0.05 mg |
€ 361.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18730714
|
Novus Biologicals
NBP1-43363-0.1mg |
0.1 mg |
€ 550.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030404
|
Novus Biologicals
NBP1-43363-0.5MG |
0.5 mg |
€ 795.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Block/Neutralize.Specifications
| M-CSF R/CD115 | |
| 0.5 mg/mL | |
| Blocking Assay, Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| P07333 | |
| 1436 | |
| The immunogen for this antibody was CD115. | |
| Primary | |
| Store at 4C. Do not freeze. |
| AFS98 | |
| Western Blot 1:100-1:2000, Flow Cytometry < 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing | |
| Monoclonal | |
| Rat | |
| Myeloid derived Suppressor Cell, Protein Kinase | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
| CSF1R | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title