Learn More
Macrophage and Histiocytoma Marker Antibody (D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-45278-0.02MG
 View more versions of this product
Product Code. 18137037
  • € 265.00 / 0.02mg
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin)
Macrophage and Histiocytoma Marker Monoclonal specifically detects Macrophage and Histiocytoma Marker in Human, Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Macrophage and Histiocytoma Marker
Monoclonal
0.2mg/mL
Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Mouse
Protein A or G purified
RUO
In Western blotting, it detects an antigen of 125kDa in human liver and 135kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells, e.g., Langerhans cells. Reportedly, its reactivity is restricted to histiocytes and macrophages.
Store at 4C.
IgG1 κ
Immunohistochemistry (Paraffin)
D11
Unconjugated
10mM PBS with 0.05% Sodium Azide
Membrane preparation from human hepatocytes
0.02 mg
Primary
Human, Mouse (Negative), Porcine (Negative), Rat (Negative)