missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage and Histiocytoma Marker Antibody (D11), Novus Biologicals™
Mouse Monoclonal Antibody
€ 265.00 - € 474.00
Specifications
| Antigen | Macrophage and Histiocytoma Marker |
|---|---|
| Clone | D11 |
| Concentration | 0.2mg/mL |
| Dilution | Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18137037
|
Novus Biologicals
NBP2-45278-0.02MG |
0.02 mg |
€ 265.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18157377
|
Novus Biologicals
NBP2-45278-0.1MG |
0.1 mg |
€ 474.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin)
Macrophage and Histiocytoma Marker Monoclonal specifically detects Macrophage and Histiocytoma Marker in Human, Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Macrophage and Histiocytoma Marker | |
| 0.2mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse (Negative), Porcine (Negative), Rat (Negative) | |
| Membrane preparation from human hepatocytes | |
| Primary | |
| Store at 4C. |
| D11 | |
| Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% Sodium Azide | |
| IgG1 κ | |
| Protein A or G purified | |
| In Western blotting, it detects an antigen of 125kDa in human liver and 135kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells, e.g., Langerhans cells. Reportedly, its reactivity is restricted to histiocytes and macrophages. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title